• USA Home
  • CRM142 - PAHs - Silty Loam Soil

EMAIL THIS PAGE TO A FRIEND
CRM142 Supelco

PAHs - Silty Loam Soil

pkg of 50 g

Purchase

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Specifications

View example COA for CRM142

Note - The link provided is an example certificate of analysis (COA) only and may not be the lot that you receive. All analytes may not be present or certified for each lot, and analyte concentrations will vary by lot.

Analyte

Description

Product #

Add to Cart

Acenaphthene    
Acenaphthylene    
Anthracene    
Benzo[b]fluoranthene    
Benzo[k]fluoranthene    
Benzo[ghi]perylene    
Benzoic acid    
Chrysene    
Dibenz[a,h]anthracene    
Fluoranthene    
Fluorene    
Phenanthrene    
Pyrene    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?