EMAIL THIS PAGE TO A FRIEND
CRM497 Supelco

pH - Sandy Clay

certified reference material, pkg of 100 g

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Environmental Matrix CRMs (RTC),
grade   certified reference material
packaging   pkg of 100 g
mfr. no.   RTC, CRM497-100
matrix   soil
Featured Industry   Environmental
format   matrix material

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Specifications

View example COA for CRM497

Note - The link provided is an example certificate of analysis (COA) only and may not be the lot that you receive. All analytes may not be present or certified for each lot, and analyte concentrations will vary by lot.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?