• USA Home
  • CRM852 - Chlordane - Sediment 1

EMAIL THIS PAGE TO A FRIEND
CRM852 Supelco

Chlordane - Sediment 1

certified reference material, pkg of 50 g

Purchase

Properties

Related Categories A-D, Additional Standards, All Herbicide Standards, Analytical Standards, Analytical/Chromatography,
grade   certified reference material
packaging   pkg of 50 g
mfr. no.   RTC, CRM852-050
concentration   235 μg/kg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
pH   6.57
Featured Industry   Agriculture
Environmental
format   matrix material

Description

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?