EMAIL THIS PAGE TO A FRIEND
D8890 Sigma-Aldrich

Dipyrone

analytical standard

Purchase

Properties

grade   analytical standard
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Biochem/physiol Actions

A non-steroidal anti-inflammatory drug that, when coadministered with morphine, potentiates its antinociceptive action and delays the development of tolerance. Dipyrone is a relatively selective inhibitor of cyclooxygenase-3 (COX-3), with lower activity against COX-1 and no activity against COX-2. Blocks PGE2-induced hyperalgesia in several models.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
PB1300000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?