• USA Home
  • DLW354236 - Corning® Collagen I Rat Tail Natural 100 mg

EMAIL THIS PAGE TO A FRIEND
DLW354236 Sigma-Aldrich

Corning® Collagen I Rat Tail Natural 100 mg

Purchase

Properties

biological source   rat tail (tendon)
assay   90% (SDS-PAGE)
form   liquid
packaging   pkg of 100 mg
application(s)   cell culture | mammalian: suitable
shipped in   wet ice
storage temp.   2-8°C
Gene Information   rat ... Col1a1(29393)

Description

General description

Corning® Collagen I, rat tail, 100 mg, is used either as a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function.

Quality

Purity = 90% by SDS-PAGE
Tested for: ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
Forms firm gel up to a dilution of 1:10; further dilution will decrease rigidity of gel
Tested and found negative for bacteria, fungi, and mycoplasma

Physical form

Liquid in 0.02 N acetic acid, concentration range 3-4 mg/ mL

Storage and Stability

Stable for at least three months at 2 °C to 8 °C. Do not freeze.

Legal Information

Corning is a registered trademark of Corning, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?