• USA Home
  • ERMDA193 - Human serum (cortisol spiked)

EMAIL THIS PAGE TO A FRIEND
ERMDA193 Sigma-Aldrich

Human serum (cortisol spiked)

ERM® certified Reference Material

Purchase

Description

Analysis Note

For more information please see:
ERMDA193

Other Notes

Sales restrictions may apply

Application

Human serum (cortisol spiked) may be used as a reference material in the analysis of cortisol present in human serum using liquid chromatography coupled with tandem mass spectrometry (LC/MS/MS).

Legal Information

ERM is a registered trademark of European Commission

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?