• USA Home
  • H161 - (±)-N-Hydroxy-3,4-methylenedioxyamphetamine

EMAIL THIS PAGE TO A FRIEND
H161 Supelco

(±)-N-Hydroxy-3,4-methylenedioxyamphetamine

analytical standard

Synonym: (±)-N-Hydroxy MDA

  • CAS Number 114562-59-3

  • Empirical Formula (Hill Notation) C10H13NO3

  • Molecular Weight 195.22

  •  MDL number MFCD00274047

  •  PubChem Substance ID 329815172

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Amphetamine, Amphetamines, Analytical Standards, Analytical/Chromatography,
Quality Level   200
grade   analytical standard
form   neat
drug control   USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IA (Portugal)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
SMILES string   CC(Cc1ccc2OCOc2c1)NO
InChI   1S/C10H13NO3/c1-7(11-12)4-8-2-3-9-10(5-8)14-6-13-9/h2-3,5,7,11-12H,4,6H2,1H3
InChI key   FNDCTJYFKOQGTL-UHFFFAOYSA-N

Description

Application

(±)-N-Hydroxy-3,4-methylenedioxyamphetamine may be used as a reference standard in the determination of N-hydroxy-3,4-methylenedioxyamphetamine in biological samples using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?