• USA Home
  • I120 - Iofetamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
I120 Supelco

Iofetamine hydrochloride

analytical standard, for drug analysis

Synonym: (±)-2-(N-Isopropyl)amino-4′-iodophenylpropane

  • CAS Number 95896-48-3

  • Empirical Formula (Hill Notation) C12H18IN · HCl

  • Molecular Weight 339.64

  •  MDL number MFCD00040805

  •  PubChem Substance ID 329815345

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Amphetamines, Analytical Standards, Analytical/Chromatography, Chemical Structure,
Quality Level   100
grade   analytical standard, for drug analysis
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
Veterinary
format   neat
SMILES string   Cl.CC(C)NC(C)Cc1ccc(I)cc1
InChI   1S/C12H18IN.ClH/c1-9(2)14-10(3)8-11-4-6-12(13)7-5-11;/h4-7,9-10,14H,8H2,1-3H3;1H
InChI key   AFLDFEASYWNJGX-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

25 mg in poly bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?