• USA Home
  • IRMM632 - Copper-65Cu solution

EMAIL THIS PAGE TO A FRIEND
IRMM632

Copper-65Cu solution

IRMM® certified Reference Material, 1 M in nitric acid

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   IRMM® certified Reference Material
mfr. no.   JRC
concentration   1 M in nitric acid
Featured Industry   Pharmaceutical (small molecule)
format   single component solution

Description

General description

Isotope abundance ratio

Analysis Note

For more information please see:
IRMM632

Legal Information

IRMM is a registered trademark of European Commission

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
EUH071
WGK Germany 
WGK 2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?