• USA Home
  • MDL015 - Sulfate, MDL Standard 10mg/L

EMAIL THIS PAGE TO A FRIEND
MDL015 Supelco

Sulfate, MDL Standard 10mg/L

certified reference material

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Elemental & Metal Standards,
grade   certified reference material
mfr. no.   RTC, MDL-015
concentration   in H2O
application(s)   ion chromatography: suitable
Featured Industry   Environmental
format   matrix material

Description

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?