EMAIL THIS PAGE TO A FRIEND
MDL019 Supelco

COD, MDL Standard

certified reference material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Biological & Chemical Oxygen Demand (BOD / COD), Certified Reference Materials (CRMs),
Quality Level   100
grade   certified reference material
mfr. no.   RTC, MDL-019
concentration   in H2O
application(s)   TOC: suitable
Featured Industry   Environmental
format   matrix material

Description

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?