• USA Home
  • MIC012 - Standard Plate Count PT- WP

EMAIL THIS PAGE TO A FRIEND
MIC012 Supelco

Standard Plate Count PT- WP

Proficiency Testing Material

  •  NACRES NA.85

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Microbiological PTs, Microbiology,
Quality Level   100
packaging   set of
mfr. no.   RTC, MIC-012
Featured Industry   Environmental
Food and Beverages
shipped in   wet ice
storage temp.   2-8°C

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Packaging

Set contains 2 sterile screw cap vials each containing a Vitroid (a small disk containing the possible organisms) and 2 - 99mL bottles of phosphate buffer.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

This PT sample is designed for the quantitative analysis heterotrophic bacteria. The PT sample is provided in duplicate as a stable dehydrated vitroid requiring hydration. The additional sample is provided for mishaps in the lab. Store the PT sample as directed. The dehydrated vitroid sample is stable for a time exceeding the study schedule.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-3 July 7 - August 20, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Legal Information

Vitroid is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Heterotrophic plate count 5.00 - 500 CFU/mL    
Heterotrophic plate count, MPN 5.00 - 500 MPN    
Safety & Documentation

Safety Information

WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?