• USA Home
  • MIC022 - E.coli in Seawater - Quantitative WP

EMAIL THIS PAGE TO A FRIEND
MIC022 Supelco

E.coli in Seawater - Quantitative WP

  •  NACRES NA.85

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Microbiological PTs, Microbiology,
Quality Level   100
packaging   pkg of 1 × 2 ea
mfr. no.   RTC, MIC-022
matrix   Vitroid
Featured Industry   Environmental
Food and Beverages
format   matrix material
shipped in   wet ice
storage temp.   2-8°C
suitability   Escherichia coli
  coliforms

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34.

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Packaging

Set contains 2 sterile screw cap vials each containing a Vitroid (a small disk containing the possible organisms) and 2 - 99mL bottles of phosphate buffer.

Analysis Note

This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005, ISO GUIDES 34 and 35, and ISO/IEC 17043.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-3 July 7 - August 20, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

This PT sample is designed for the quantitative analysis of total and/or fecal coliform and E. coli in seawater. The PT sample is provided in duplicate as a stable dehydrated vitroid requiring hydration. The additional sample is provided for mishaps in the lab. Store the PT sample as directed. The dehydrated vitroid sample is stable for a time exceeding the study schedule.

Legal Information

Vitroid is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Total coliforms 20.0 - 2400 CFU/100mL    
Total Coliform, MPN 20.0 - 2400 MPN/100 mL    
Escherichia coli 20.0 - 2400 CFU/100mL    
Escherichia coli, MPN 20.0 - 2400 MPN/100 mL    
Fecal coliforms 20.0 - 2400 CFU/100mL    
Fecal coliform, MPN 20.0 - 2400 MPN/100 mL    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?