EMAIL THIS PAGE TO A FRIEND
NIST17F

Sucrose

NIST® SRM® 17f, optical rotation

Synonym: α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

  • CAS Number 57-50-1

  • Empirical Formula (Hill Notation) C12H22O11

  • Molecular Weight 342.30

  •  Beilstein/REAXYS Number 90825

  •  MDL number MFCD00006626

  •  PubChem Substance ID 329818715

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, NIST Standards, NIST Standards S - T, Pharmacopeia & Metrological Institutes Standards More...
Quality Level   100
packaging   pkg of 60 g
mfr. no.   NIST
mp   185-187 °C (lit.)
Featured Industry   Pharmaceutical (small molecule)
format   neat
SMILES string   OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
InChI   1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1
InChI key   CZMRCDWAGMRECN-UGDNZRGBSA-N

Description

General description

SRM 17F_cert SRM 17F _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

SRM is a registered trademark of National Institute of Standards and Technology

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?