• USA Home
  • PE10025 - Chlorine in Water - Field PT

EMAIL THIS PAGE TO A FRIEND
PE10025 Supelco

Chlorine in Water - Field PT

Proficiency Testing Material

  •  NACRES NA.24

Purchase

Description

Other Notes

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Total Residual Chlorine 0.00 - 1.00 mg/L    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?