EMAIL THIS PAGE TO A FRIEND
PE1052 Supelco

Trace Metals 2 - WP

Proficiency Testing Material, Concentrate for dilution to 1L

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, DMRQA PTs, Proficiency Testing,
Quality Level   100
packaging   ampule of 1 × 20 mL
mfr. no.   RTC, PEI-034-2
concentration   in nitric acid
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is packaged as a 20mL concentrate. The solvent for this sample is 5% nitric acid by volume in DI water. The PT sample has not been preserved.
Sample Preparation
To prepare solution, partially fill a 1000mL volumetric flask with reagent-grade water (near 20ºC) and add 1mL of trace-metal-grade nitric acid.
Shake ampule well prior to opening.
Open ampule PE1052-20ML (be careful to avoid cuts on glass) and transfer 10.0mL of concentrate to the volumetric flask.
Fill the flask to volume with reagent-grade water and mix well.
The sample is now ready for immediate analysis.
A reagent-grade blank should be analyzed concurrently for background correction. The water blank should be made to match the acid matrix of the standards and samples.
Each analyte is reported in ug/L.
Note - The 0.15% v/v nitric acid specification is a minimum; the amount and type of acid may be altered to routine analytical procedures if needed.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-2 March 24 - May 7, 2021
WP21-3 July 7 - August 20, 2021
WP21-4 September 22 - November 5, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Antimony, Sb 90-900 μg/L    
Barium, Ba 100-2500 μg/L    
Boron, B 800-2000 μg/L    
Molybdenum, Mo 60-600 μg/L    
Silver, Ag 100-1000 μg/L    
Strontium, Sr 50-500 μg/L    
Thallium, Tl 80-800 μg/L    
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

QC1052 Trace Metals 2-WP, certified reference material
94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?