• USA Home
  • PE1091 - Volatile Residue - WP

EMAIL THIS PAGE TO A FRIEND
PE1091 Supelco

Volatile Residue - WP

Proficiency Testing Material, Concentrate for dilution to 1L

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Pollution/Waste Water Inorganics,
Quality Level   100
packaging   ampule of 1 × 1.2 g
mfr. no.   RTC, PEI-270
matrix   NaCl
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is as a concentrate for dilution in DI water.
The solvent for this sample is NaCl.
The sample must be stored at room temperature.

Sample Preparation

Partially fill a 1000mL volumetric flask with reagent-grade water.
Open vial PE1091-1.2G and weigh 500mg of solids and quantitatively transfer to the volumetric flask using a glass funnel.
Rinse the flask with reagent-grade water.
Fill the volumetric flask to volume (1000mL) and mix by inverting the flask 10 times.
Let the flask stand for 10 minutes and invert again 10 times.
The sample is now ready for immediate analysis.
Do not filter sample prior to volatile analysis. Analyte losses may occur.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-2 March 24 - May 7, 2021
WP21-3 July 7 - August 20, 2021
WP21-4 September 22 - November 5, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Volatile Solids, Total 100-500 mg/L    
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

QC1091 Volatile Residue - WP, certified reference material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?