• USA Home
  • PE1116 - PCB Congeners in Water - WP

EMAIL THIS PAGE TO A FRIEND
PE1116 Supelco

PCB Congeners in Water - WP

Proficiency Testing Material

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Organic Pollutant Standards, PCBs & Dioxins Standards,
Quality Level   100
packaging   ampule of 1 × 2 mL
mfr. no.   RTC, PEO-403
concentration   in acetone
Featured Industry   Environmental
storage temp.   2-8°C

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is packaged as a 1.5 mL concentrate.
The PT sample is not preserved.
The sample must be stored at 4 °C.
Expected concentration of each congener is 1 to 10 ng/L.

Sample preparation
Allow the ampule concentrate to equilibrate to room temperature.
Fill a 1000 mL Class A Volumetric Flask with 900 mL of organic free reagent water.
Open the ampule and pipet exactly 1.0 mL of the concentrate into the flask.
Fill to volume and mix well.
This represents the sample for analysis.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released unti the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-3 July 7 - August 20, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

PCBs, total 1-250 ng/L    
PCB (20)+(28) 1-40 ng/L    
2,2',5,5'-Tetrachlorobiphenyl (PCB 52) 1-10 ng/L    
3,3',4,4'-Tetrachlorobiphenyl (PCB 77) 1-10 ng/L    
3,4,4',5-Tetrachlorobiphenyl (PCB 81) 1-10 ng/L    
2,2',4,5,5'-Pentachlorobiphenyl (PCB 101) 1-10 ng/L    
PCB (90)+(101)+(113) 1-50 ng/L    
2,3,3',4,4'-Pentachlorobiphenyl (PCB 105) 1-10 ng/L    
2,3',4,4',5-Pentachlorobiphenyl (PCB 118) 1-10 ng/L    
2,3',4,4',5'-Pentachlorobiphenyl (PCB 123) 1-10 ng/L    
2,3,4,4',5-Pentachlorobiphenyl (PCB 114) 1-10 ng/L    
3,3',4,4',5-Pentachlorobiphenyl (PCB 126) 1-10 ng/L    
2,2',3,4,4',5'-Hexachlorobiphenyl (PCB 138) 1-10 ng/L    
PCB (129)+(138)+(163) 1-50 ng/L    
PCB (153)+(168) 1-50 ng/L    
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB 153) 1-10 ng/L    
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB 157) 1-10 ng/L    
PCB (156)+(157) 1-50 ng/L    
2,3,3',4,4',5-Hexachlorobiphenyl (PCB 156) 1-10 ng/L    
2,3',4,4',5,5'-Hexachlorobiphenyl (PCB 167) 1-10 ng/L    
3,3',4,4',5,5'-Hexachlorobiphenyl (PCB 169) 1-10 ng/L    
PCB (180)+(193) 1-50 ng/L    
2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB 189) 1-10 ng/L    
2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB 180) 1-10 ng/L    
2,4,4'-Trichlorobiphenyl (PCB 28) 1-10 ng/L    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
Supplemental Hazard Statements 
EUH066
RIDADR 
UN1090 - class 3 - PG 2 - Acetone, solution
WGK Germany 
WGK 1
Flash Point(F) 
1.4 °F - closed cup - Solvent
Flash Point(C) 
-17 °C - closed cup - Solvent

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?