• USA Home
  • PE1212 - Triazine Pesticides - WP

EMAIL THIS PAGE TO A FRIEND
PE1212 Supelco

Triazine Pesticides - WP

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Pollution/Waste Water Organics,
Quality Level   100
mfr. no.   RTC, PEO-122-4
concentration   in acetone
Featured Industry   Environmental
storage temp.   2-8°C

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

Description
This PT sample is packaged as a 1.5 mL concentrate.
The solvent for this sample is acetone.
The PT sample is not preserved.

Sample Preparation- PLEASE REFER TO YOUR INSTRUCTION PACKET FOR LATEST INSTRUCTIONS
1) Fill a 1000 mL Class A Volumetric Flask with 950 mL of organic free reagent water.
2) Allow the amule concentrate to equilibrate to room temperature.
3) Open the ampule PE1212-2ML and pipet exactly 1.0 mL of the concentrate into the flask.
4) Bring the flask to volume with reagent water.
5) Stopper the flask and mix well.
6) This represents the sample for analysis.

Other Notes

Concentrations may vary and not all analytes may be present

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2019-2020)

Wastewater
WP19-1 January 16 - March 1, 2019
WP19-2 March 27 - May 10, 2019
WP19-3A May 15 - June 28, 2019
WP19-3B July 10 - August 23, 2019
WP19-4 September 25 - November 8, 2019
WP19-5 November 20 - January 3, 2020

WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Ametryn 1 - 15 ug/L    
Atraton 1 - 15 ug/L    
Atrazine 1 - 15 ug/L    
Carbatamide 0 - 10 µg/L    
Cyanazine 1 - 15 ug/L    
Cyfluthrin 0 - 10 µg/L    
Deisopropylatrazine 1 - 15 ug/L    
Diaminoatrazine 1 - 15 ug/L    
Diflufenican 0 - 10 µg/L    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
Supplemental Hazard Statements 
EUH066
RIDADR 
UN1090 - class 3 - PG 2 - Acetone, solution
WGK Germany 
WGK 1
Flash Point(F) 
1.4 °F - closed cup - Solvent
Flash Point(C) 
-17 °C - closed cup - Solvent

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?