EMAIL THIS PAGE TO A FRIEND
PE1274 Supelco

Acid Compounds - WP

Proficiency Testing Material, Concentrate for dilution to 1L

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Pollution/Waste Water Organics,
Quality Level   100
packaging   ampule of 1.2 mL
mfr. no.   RTC, PEO-022
concentration   in acetone
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

Randomly contains a minimum 80% of the analytes listed.

This PT sample is packaged as a 1.2 mL concentrate.
The solvent for this sample is acetone.
The PT sample is not preserved.
MMW-001 is intended to modify the products to simulate a waste water sample. The final dilution will replace the use of DI (Reagent Grade) water for the prepartion of the appropriate CRM/QC sample. The sample will add turbidity and TSS to the final volume. These do not affect the properties of the analytes of interest in the final sample if prepared according to instructions. Note filtration may be necessary as required by your method.

To use the matrix modifier simply mix Waste Water Matrix Modifier as specified in the MMW-001 instructions. Use this solution in place of the reagent grade water in the instructions above.

Sample preparation
To prepare solution, add 1L of reagent water to a 2L separatory funnel.
Open ampule PE1274-1.2ML (be careful to avoid cuts on glass) and transfer exactly 1.0 mL of the concentrate to the searatory funnel.
Mix well.
The sample is now ready for analysis.
For calculations, assume initial sample volume was 1000 mL.

The packsize on this sample has recently changed from 2ML to 1.2ML. This change does not affect the performance of the proficiency test.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released unti the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-2 March 24 - May 7, 2021
WP21-3 July 7 - August 20, 2021
WP21-4 September 22 - November 5, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Benzoic acid 50-200 μg/L    
Benzoic acid 50-200 μg/L    
4-Chloro-3-methylphenol 30-200 μg/L    
2-Chlorophenol 30-200 μg/L    
2,4-Dichlorophenol 40-190 μg/L    
2,6-Dichlorophenol 40-200 μg/L    
2,4-Dimethylphenol 65-200 μg/L    
3,5-Dinitrophenol 20-200 μg/L    
2,4-Dinitrophenol 0-180 μg/L    
2-Methyl-4,6-dinitrophenol 60-200 μg/L    
2-Methylphenol (o-Cresol) 1 50-200 μg/L    
3-Methylphenol (m-Cresol) 50-200 μg/L    
4-Methylphenol (p-Cresol) 50-200 μg/L    
3+4-Methylphenol (m+p-Cresol) 50-200 μg/L    
2-Nitrophenol 1 50-190 μg/L    
3-Nitrophenol 50-200 μg/L    
4-Nitrophenol 100-180 μg/L    
Pentachlorophenol 55-200 μg/L    
Phenol 100-200 μg/L    
2,3,4,6-Tetrachlorophenol 50-200 μg/L    
2,4,5-Trichlorophenol 50-200 μg/L    
2,4,6-Trichlorophenol 50-200 μg/L    
2,5-Dimethylphenol 20-200 μg/L    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Central nervous system
Supplemental Hazard Statements 
EUH066
RIDADR 
UN1090 - class 3 - PG 2 - Acetone, solution
WGK Germany 
WGK 2
Flash Point(F) 
1.4 °F - closed cup - Solvent
Flash Point(C) 
-17 °C - closed cup - Solvent

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

QC1274 Acids - WP, certified reference material
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?