• USA Home
  • PE1282 - Volatile Residue/Fixed Solids - WP

EMAIL THIS PAGE TO A FRIEND
PE1282 Supelco

Volatile Residue/Fixed Solids - WP

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Pollution/Waste Water Inorganics,
Quality Level   100
packaging   ampule of 1 × 1.2 g
mfr. no.   RTC, PEI-270-2
matrix   NaCl
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is as a concentrate for dilution in DI water.
The sample must be stored at room temperature.

Sample Preparation

Partially fill a 1000mL volumetric flask with reagent-grade water.
Open vial PE1282-1.2G and weigh 500mg of solids and quantitatively transfer to the volumetric flask using a glass funnel.
Rinse the flask with reagent-grade water.
Fill the volumetric flask to volume (1000mL) and mix by inverting the flask 10 times.
Let the flask stand for 10 minutes and invert again 10 times.
The sample is now ready for immediate analysis.
“Fixed Solids” refers to the portion of the sample remaining after the ashing process (550ºC). “Volatile Residue” refers to the amount of sample lost during the ashing process (550ºC).

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-3 July 7 - August 20, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Solids, Fixed 50-1000 mg/L    
Volatile Solids, Total 100-500 mg/L    
Volatile Suspended Solids 0.00 - 500 mg/L    
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?