• USA Home
  • PE1295 - Dioxins in Water - WP

EMAIL THIS PAGE TO A FRIEND
PE1295 Supelco

Dioxins in Water - WP

Proficiency Testing Material

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Pollution/Waste Water Organics,
Quality Level   100
packaging   ampule of 1 × 2 mL
mfr. no.   RTC, PEO-258
concentration   in toluene
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This concentrate contains up to 16 dioxins and furans in toluene

Sample preparation
Pour 1000-mL of reagent grade DI water into a extraction vessel.
Open ampule and transfer 1 mL of concentrate to the extraction vessel using Class A glassware.
Analyze as required by your normal procedures.
Assume a 1-Liter sample volume for calculation purposes.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released unti the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Wastewater
WP20-1 January 15 - February 28, 2020
WP20-2 March 25 - May 8, 2020
WP20-3A May 13 - June 26, 2020
WP20-3B July 8 - August 21, 2020
WP20-4 September 23 - November 6, 2020
WP20-5 November 25 - January 8, 2021

WP21-1 January 13 - February 26, 2021
WP21-3 July 7 - August 20, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

1,2,3,4,6,7,8-Hpcdf 50 - 2000 pg/L    
1,2,3,4,7,8,9-Hpcdf 50 - 2000 pg/L    
1,2,3,4,6,7,8-Hpcdd 50 - 2000 pg/L    
Total Heptachlorodibenzo-p-dioxin (Total HPCDD) 50 - 4000 pg/L    
Total Heptachlorodibenzofuran (Total HPCDF) 50 - 4000 pg/L    
1,2,3,4,7,8-Hxcdd 50 - 2000 pg/L    
1,2,3,6,7,8-Hxcdd 50 - 2000 pg/L    
1,2,3,7,8,9-Hxcdd 50 - 2000 pg/L    
Hxcdd, total 50 - 6000 pg/L    
1,2,3,4,7,8-Hxcdf 50 - 2000 pg/L    
1,2,3,6,7,8-Hxcdf 50 - 2000 pg/L    
1,2,3,7,8,9-Hxcdf 50 - 2000 pg/L    
2,3,4,6,7,8-Hxcdf 50 - 2000 pg/L    
Total Hexachlorodibenzofuran (Total HxCDF) 50 - 8000 pg/L    
1,2,3,4,6,7,8,9-OCDF 50 - 2000 pg/L    
1,2,3,4,6,7,8,9-OCDD 50 - 2000 pg/L    
1,2,3,7,8-Pecdd 50 - 2000 pg/L    
1,2,3,7,8-Pecdf 50 - 2000 pg/L    
2,3,4,7,8-Pecdf 50 - 2000 pg/L    
Pecdf, total 50 - 6000 pg/L    
Pecdd, total 50 - 2000 pg/L    
TCDD, total 50 - 10000 pg/L    
2,3,7,8-TCDF 50 - 2000 pg/L    
TCDF, total 50 - 10000 pg/L    
2,3,7,8-TCDD 50 - 2000 pg/L    
PCDF, total 50 - 10000 pg/L    
PCDD, total 50 - 10000 pg/L    
Total Polychlorinated dibenzo-p-dioxins and total Polychlorinated dibenzofurans (Total PCDD/Fs) 1 50 - 20000 pg/L    
Safety & Documentation

Safety Information

Signal word 
Danger
Target organs 
Central nervous system
RIDADR 
UN1993 - class 3 - PG 2 - EHS - VP <= 110kPa - Flammable liquids, n.o.s., HI: all
WGK Germany 
WGK 3
Flash Point(F) 
39.2 °F - closed cup - Solvent
Flash Point(C) 
4 °C - closed cup - Solvent

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

QC1295 Dioxin and Furans in Water - WP, certified reference material
94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?