• USA Home
  • PE1361 - Bromate and Bromide - WS

EMAIL THIS PAGE TO A FRIEND
PE1361 Supelco

Bromate and Bromide - WS

Proficiency Testing Material

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Supply/Drinking Water Inorganics,
Quality Level   100
packaging   ampule of 1 × 2 mL
mfr. no.   RTC, PEI-017-1
concentration   in H2O
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This ampule contains a concentrate for preparation of a solution that can be analyzed for bromate and bromide.
This PT sample is packaged as a 2 mL concentrate.
The solvent for this sample is water.
The PT sample has been autoclaved for preservation.
The sample must be stored at 4 degrees Celsius.

Sample preparation
Partially fill a 1000-mL volumetric flask with reagent-grade water (near 20 degrees C).
Open ampule PE1361 (be careful to avoid cuts on glass) and transfer 1.0 mL of concentrate to the volumetric flask.
Fill the volumetric flask to volume with reagent-grade water and mix well.
The sample is now ready for immediate analysis.
The sample should be analyzed as soon as possible after opening the ampule and diluting to volume.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Drinking Water
WS20-1 January 15 - February 28, 2020
WS20-2 March 25 - May 8, 2020
WS20-3 July 8 - August 21, 2020
WS20-4 September 23 - November 6, 2020

WS21-1 January 13 - February 26, 2021
WS21-2 March 24 - May 7, 2021
WS21-3 July 7 - August 20, 2021
WS21-4 September 22 - November 5, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Bromate 7-50 μg/L    
Bromide 50-300 μg/L    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

QC1361 Bromate and Bromide-WS, certified reference material
94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?