• USA Home
  • PE1380 - Formaldehyde in Water - PT

EMAIL THIS PAGE TO A FRIEND
PE1380 Supelco

Formaldehyde in Water - PT

Proficiency Testing Material

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Pollution/Waste Water Organics,
Quality Level   100
packaging   ampule of 20 mL
mfr. no.   RTC, PEO-417
concentration   in H2O
Featured Industry   Environmental
storage temp.   2-8°C

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is supplied as a 20 mL concentrate.
The solvent for this sample is water.

Sample preparation

To prepare solution, partially fill a 1000-mL volumetric flask with ammonia-free, reagent-grade water (near 20 degrees C).
Open ampule PEO-417 (be careful to avoid cuts on glass) and transfer 10.0 mL of concentrate to the volumetric flask.
Fill the volumetric flask to volume with reagent-grade water and mix well.
The sample is now ready for immediate analysis.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released unti the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Drinking Water
WS20-1 January 15 - February 28, 2020
WS20-2 March 25 - May 8, 2020
WS20-3 July 8 - August 21, 2020
WS20-4 September 23 - November 6, 2020

WS21-1 January 13 - February 26, 2021
WS21-3 July 7 - August 20, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

Formaldehyde 5-500 mg/L    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

QC1380 Formaldehyde in Water
94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?