• USA Home
  • PE1398 - PBDE′s in water - PT

EMAIL THIS PAGE TO A FRIEND
PE1398 Supelco

PBDE′s in water - PT

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Water Pollution/Waste Water Organics,
Quality Level   100
packaging   ampule of 1 × 2 mL
mfr. no.   RTC, PEO-415
concentration   in acetone
Featured Industry   Environmental
storage temp.   2-8°C

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample


This PT is valid for ISO method 22032.

Preparation Note

This PT sample is packaged as a 1.5 mL concentrate.
The PT sample is not preserved.
The sample must be stored at 4 °C

Sample preparation
Allow the ampule concentrate to equilibrate to room temperature.
Fill a 1000 mL Class A Volumetric Flask with 900 mL of organic free reagent water.
Open the ampule and pipet exactly 1.0 mL of the concentrate into the flask.
Fill to volume and mix well.
This represents the sample for analysis.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released unti the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2020-2021)

Drinking Water
WS20-1 January 15 - February 28, 2020
WS20-2 March 25 - May 8, 2020
WS20-3 July 8 - August 21, 2020
WS20-4 September 23 - November 6, 2020

WS21-1 January 13 - February 26, 2021
WS21-3 July 7 - August 20, 2021

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

BDE 153 20-500 ng/L    
BDE 99 20-500 ng/L    
BDE 100 20-500 ng/L    
BDE 47 20-500 ng/L    
BDE 154 20-500 ng/L    
BDE 183 20-500 ng/L    
BDE 205 20-500 ng/L    
BDE 209 20-500 ng/L    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
Supplemental Hazard Statements 
EUH066
RIDADR 
UN1090 - class 3 - PG 2 - Acetone, solution
WGK Germany 
WGK 1
Flash Point(F) 
1.4 °F - closed cup - Solvent
Flash Point(C) 
-17 °C - closed cup - Solvent

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?