• USA Home
  • PE1799 - TPH in Water (low level)

EMAIL THIS PAGE TO A FRIEND
PE1799 Supelco

TPH in Water (low level)

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Underground Storage Tank PTs (UST),
packaging   ampule of 1 × 2 mL
mfr. no.   RTC, PEO-115TX-L
concentration   in pentane
Featured Industry   Environmental
storage temp.   2-8°C

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released unti the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2019-2020)

Soils
LPTP19-S1 January 30 - March 15, 2019
LPTP19-S2 April 10 - May 24, 2019
LPTP19-S3 July 24 - September 6, 2019
LPTP19-S4 October 9 - November 22, 2019

LPTP20-S1 January 29 - March 13, 2020
LPTP20-S2 April 8 - May 22, 2020
LPTP20-S3 July 22 - September 4, 2020
LPTP20-S4 October 7 - November 20, 2020

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Preparation Note

This vial contains a concentrate of a blended gasoline from three different sources and diesel fuel oil No. 2 in pentane for analysis by TNRCC Method 1005.
When prepared according to instructions, the concentration of each analyte will be less than 10 mg/L.

Sample preparation
To prepare solution, fill a 40-mL VOA vial to volume with carbon-filtered reagent-grade water at room temperature.
Remove enough water to allow for the addition of extraction solvent.
Open vial PE1799-2ML and immediately withdraw 40 μL with a gas tight syringe.
Quickly inject the 40 μL aliquot below the surface of the reagent water in the voa vial.
Add extraction solvent.
Mix the sample by inverting the flask three times only (excessive shaking results in loss of volatiles) and analyze immediately. Assume a 40 mL sample for calculation purposes. Note this is a 1:1000 dilution and other dilutions are acceptable.

Analyte

Description

Product #

Add to Cart

C6 Aliphatics 0.000 - 10.0 mg/L    
>C7-C8 Aromatics 0.000 - 10.0 mg/L    
Residual Range Organics (RRO),>C28-C35 0.000 - 10.0 mg/L    
Total Petroleum Hydrocarbons (TPH), (C6-C35 5-50 mg/L    
>C6 to C8 Aliphatics 0.000 - 10.0 mg/L    
>C8 to C10 Aliphatics 0.000 - 10.0 mg/L    
>C10 to C12 Aliphatics 0.000 - 10.0 mg/L    
>C12 to C16 Aliphatics 0.000 - 10.0 mg/L    
>C16 to C21 Aliphatics 0.000 - 10.0 mg/L    
>C21 to C35 Aliphatics 0.000 - 10.0 mg/L    
>C8 to C10 Aromatics 0.000 - 10.0 mg/L    
>C10 to C12 Aromatics 0.000 - 10.0 mg/L    
>C12 to C16 Aromatics 0.000 - 10.0 mg/L    
>C16 to C21 Aromatics 0.000 - 10.0 mg/L    
>C21 to C35 Aromatics 0.000 - 10.0 mg/L    
Diesel Range Organics (DRO) 0.000 - 10.0 mg/L    
Diesel-range total petroleum hydrocarbons, >C12-C28 0.000 - 10.0 mg/L    
Gasoline range organics (GRO), C5-C10 0.000 - 10.0 mg/L    
Gasoline Range Organics, C6-C12 0.000 - 10.0 mg/L    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 2 - Pentanes
WGK Germany 
3
Flash Point(F) 
-56.2 °F
Flash Point(C) 
-49 °C

Documents

Certificate of Analysis

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products.Once you log onto our website, you will find the price and availability displayed on the product detail page.You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 orview local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Are Certificates of Analysis created forRTC proficiency testing products?
Certificates of Analysis are not available for RTC proficiency testing products. Those are the samples with the SPE or PE prefix in the part number.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?