EMAIL THIS PAGE TO A FRIEND
PFS14 Supelco

SpheriCal Mix

Protein Low, suitable for MALDI-MS

  •  NACRES NA.21

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, MS Standards, Physical Properties Standards, Single Compounds More...
Quality Level   100
shelf life   expiry date on the label
mol wt   Mr 7500-15000 Da (Na & adduct)
composition   9-nitroanthracene, >90%
  sodium trifluoroacetate, <10%
application(s)   MALDI-MS: suitable
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
storage temp.   −20°C

Description

General description

SpheriCal is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibration spots, SpheriCal offers reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.

Mass ranges available:

i. SpheriCal Mix Peptide Low (m/z 500 - 1,600)
ii. SpheriCal Mix Peptide Medium (m/z 1,600 - 3,500)
iii. SpheriCal Mix Peptide High (m/z 3,500 - 7,500)
iv. SpheriCalMix Protein Low (m/z 7,500 - 15,000)
v. SpheriCalMix Protein Medium (m/z 15,000 - 30,000)

SpheriCal Mix: quick and easy to use - just add solvent.

As calibration standard, matrix and counter ion are pre-mixed in optimized ratios, SpheriCal Mix dramatically expedites sample preparation for calibration in minutes. SpheriCal Mix 9NA is pre-formulated with 9-nitroanthracene matrix for a robust calibration solution.

Find more information here SpheriCal

calibrants PFS4A : PFS4B : PFS4C : PFS4D = 1:1:1:1
Matrix/counter ion: 9-Nitroanthracene/Na+

Application

For mass calibration in mass spectrometry

Packaging

Quantity: 3*5 mg

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Grayson Group - Professor Product Portal

From our library of Articles, Sigma-Aldrich presents Grayson Group - Professor Product Portal
Keywords: Coupling reactions, Mass spectrometry, Nuclear magnetic resonance spectroscopy, Phase transitions, Polymerization reactions, Solvents

MALDI-TOF Characterization of Functionalized Polymers

With an increased demand for high performance and specialized polymeric materials, tools for selective end group functionalization and modification have been a focus of synthetic development. Due to ...
Molly E. Payne and Scott M. Grayson
Department of Chemistry, Tulane University, New Orleans, Louisiana
Keywords: Amidations, Bacterial conjugations, Catalysis, Coupling reactions, Cycloadditions, Ene reaction, Mass spectrometry, Nuclear magnetic resonance spectroscopy, PEGylations, Peptide synthesis, Phase transitions, Polymerization reactions, Transesterifications

SpheriCal - Monodisperse Dendrimers as Universal Calibrants for MALDI-Mass Spectrometry

About SpheriCal SpheriCal is a series of monodisperse calibration standards that offer mass spectrometry users time-saving sample preparation plus highly accurate calibration. By combining excellent ...
Keywords: Catalysis, Click chemistry, Mass spectrometry, Nanotechnology, Proteomics, Sample preparations, Semiconductor, Solvents

Related Products

related product

Product #

Description

Add to Cart

900666 Bruker standard solvent, suitable for MALDI-MS, 50% ACN + 47.5% H2O + 2.5% TFA
P2613 ProteoMass P14R MALDI-MS Standard, vial of 10 nmol
C8857 ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
C8982 α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
A8346 ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da
I6154 ProteoMass Insulin chain B oxidized MALDI-MS Standard, vial of 10 nmol
I6279 ProteoMass Insulin MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da
A8471 ProteoMass Albumin MALDI-MS Standard, vial of 10 nmol, average mol wt 66,429.09 Da by calculation
A9096 ProteoMass Aldolase MALDI-MS Standard, vial of 10 nmol, average mol wt 39,211.28 Da by calculation
A8846 ProteoMass Angiotensin II MALDI-MS Standard, vial of 10 nmol
A8971 ProteoMass Apomyoglobin MALDI-MS Standard, vial of 10 nmol, (M+H+) 16,952.27 Da by calculation
B4181 ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 756.3997 Da
MSCAL1 ProteoMass Peptide and Protein MALDI-MS Calibration Kit, Peptide and Protein MALDI MS Calibration Standard
MSCAL2 ProteoMass Peptide MALDI-MS Calibration Kit, Peptide MALDI MS Calibration Standard
MSCAL3 ProteoMass Protein MALDI-MS Calibration Kit, Protein MALDI MS Calibration Standard
56197 3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
54793 2-(4-Hydroxyphenylazo)benzoic acid, matrix substance for MALDI-MS, ≥99.5%
85429 Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
03565 MALDI validation set polystyrene Mp ~700 - 70 000, certified reference material
37468 2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%
91928 2′,4′,6′-Trihydroxyacetophenone monohydrate, matrix substance for MALDI-MS, ≥99.5%
03597 MALDI validation set PS, PMMA, PDMS, PEG and PSS Mp ~4′300 - 6′500
87884 trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, matrix substance for MALDI-MS, ≥99.0% (HPLC)
60018 Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
29208 (N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide, for protein sequence analysis (by MALDI-MS), ≥98.5%
46278 trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
85707 2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
70990 α-Cyano-4-hydroxycinnamic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
N3786 α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
39468 α-Cyano-4-hydroxycinnamic acid, matrix substance for MALDI-MS, Ultra pure
39319 2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
49508 Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
50862 Super-DHB, suitable for matrix substance for MALDI-MS, ≥99.0%
02241 2′,3′-Dideoxycytidine 5′-triphosphate sodium salt solution, suitable for MALDI-MS, ≥98.0% (HPLC)
MSCAL4 ProteoMass MALDI Calibration Kit for Exactive Series MS, LTQ XL and LTQ Hybrids, MALDI MS Calibration Standard
92817 9-Aminoacridine, matrix substance for MALDI-MS, ≥99.5% (HPLC)
56451 1,5-Diaminonaphthalene, matrix substance for MALDI-MS, ≥99.0% (HPLC)
50757 Micro-Array for Mass Spectrometry (MAMS), for single cell analysis
94141 4-Chloro-α-cyanocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
77646 α-Cyano-2,4-difluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
89063 4-Bromo-α-cyanocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
77081 α-Cyano-4-fluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
38419 α-Cyano-2,3,4,5,6-pentafluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
39379 4-Chloro-α-cyanocinnamic acid - α-Cyano-2,4-difluorocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of both components, HPLC)
68914 4-Bromo-α-cyanocinnamic acid - 4-Chloro-α-cyanocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of both components, HPLC)
03841 α-Cyano-4-hydroxycinnamic acid - α-Cyano-2,4-difluorocinnamic acid - α-Cyano-2,3,4,5,6-pentafluorocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of three components, HPLC)
05851 4-Aminoquinaldine, matrix substance for MALDI-MS, ≥99.0% (HPLC)
78246 Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
04973 Micro-Array for Mass Spectrometry (MAMS), for quantitative MALDI
94477 (E)-2-Cyano-3-(2-naphthyl)acrylic acid, matrix substance for MALDI-MS, ≥98.0% (HPLC)
83788 (2E)-3-(9-Anthryl)-2-cyanoacrylic acid, matrix substance for MALDI-MS, ≥97.0% (HPLC)
76884 Anthranilamide, matrix substance for MALDI-MS, ≥99.0% (HPLC)
56229 9-Nitroanthracene, matrix substance for MALDI-MS, ≥98.5% (HPLC)
PFS11 SpheriCal Mix, Peptide Low, suitable for MALDI-MS
PFS12 SpheriCal Mix, Peptide Medium, suitable for MALDI-MS
PFS13 SpheriCal Mix, Peptide High, suitable for MALDI-MS
PFS15 SpheriCal Mix, Protein Medium, suitable for MALDI-MS
PFS31 SpheriCal Aqua, Peptide Medium, suitable for MALDI-MS
PFS32 SpheriCal Aqua, Peptide High, suitable for MALDI-MS
PFS33 SpheriCal Aqua, Protein Low, suitable for MALDI-MS
PFS20 SpheriCal Neat, Peptide Low, suitable for MALDI-MS
PFS21 SpheriCal Neat, Peptide Medium, suitable for MALDI-MS
PFS22 SpheriCal Neat, Peptide High, suitable for MALDI-MS
PFS23 SpheriCal Neat, Protein Low, suitable for MALDI-MS
PFS24 SpheriCal Neat, Protein Medium, suitable for MALDI-MS
PFS30 SpheriCal Aqua, Peptide Low, suitable for MALDI-MS
69028 4-Phenyl-α-cyanocinnamamide, matrix substance for MALDI-MS, ≥98.5% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?