EMAIL THIS PAGE TO A FRIEND
PFS33 Supelco

SpheriCal Aqua

Protein Low, for MALDI MS

  •  NACRES NA.21

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, MS Standards, Physical Properties Standards, Single Compounds More...
Quality Level   100
grade   for MALDI MS
shelf life   expiry date on the label
mol wt   Mr 5000-10000 Da
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
storage temp.   −20°C

Description

General description

SpheriCal is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibrations, SpheriCal offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.

Mass ranges available:

i. SpheriCal Aqua peptide low (m/z 500 - 1,000)
ii. SpheriCal Aqua peptide medium (m/z 1,000 - 2,500)
iii. SpheriCal Aqua peptide high (m/z 2,500 - 5,200)
iv. SpheriCal Aqua protein low (m/z 5,200 - 11,000)

SpheriCal Aqua Neat: ideal for water-based systems.

SpheriCal Aqua is ideal for calibration with aqueous systems commonly used in life sciences, e.g., matrices DHB, HCCA and SA or solvents such as water and acetonitrile, although ionization is preferred with alkali cations. Each vial of SpheriCal Neat contains sufficient calibration standard for over 100 calibration spots.

Find more information here SpheriCal

dendritic calibrants PFQ4A : PFQ4B : PFQ4C : PFQ4D = 1:1:1:1

Application

For mass calibration in mass spectrometry

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Articles

Grayson Group - Professor Product Portal

From our library of Articles, Sigma-Aldrich presents Grayson Group - Professor Product Portal
Keywords: Coupling reactions, Mass spectrometry, Nuclear magnetic resonance spectroscopy, Phase transitions, Polymerization reactions, Solvents

MALDI-TOF Characterization of Functionalized Polymers

With an increased demand for high performance and specialized polymeric materials, tools for selective end group functionalization and modification have been a focus of synthetic development. Due to ...
Molly E. Payne and Scott M. Grayson
Department of Chemistry, Tulane University, New Orleans, Louisiana
Keywords: Amidations, Bacterial conjugations, Catalysis, Coupling reactions, Cycloadditions, Ene reaction, Mass spectrometry, Nuclear magnetic resonance spectroscopy, PEGylations, Peptide synthesis, Phase transitions, Polymerization reactions, Transesterifications

SpheriCal - Monodisperse Dendrimers as Universal Calibrants for MALDI-Mass Spectrometry

About SpheriCal SpheriCal is a series of monodisperse calibration standards that offer mass spectrometry users time-saving sample preparation plus highly accurate calibration. By combining excellent ...
Keywords: Catalysis, Click chemistry, Mass spectrometry, Nanotechnology, Proteomics, Sample preparations, Semiconductor, Solvents

Related Products

related product

Product #

Description

Add to Cart

900666 Bruker standard solvent, for MALDI MS, 50% ACN + 47.5% H2O + 2.5% TFA
P2613 ProteoMass P14R MALDI-MS Standard, vial of 10 nmol
C8857 ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
C8982 α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
A8346 ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da
I6154 ProteoMass Insulin chain B oxidized MALDI-MS Standard, vial of 10 nmol
I6279 ProteoMass Insulin MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da
A8471 ProteoMass Albumin MALDI-MS Standard, vial of 10 nmol, average mol wt 66,429.09 Da by calculation
A9096 ProteoMass Aldolase MALDI-MS Standard, vial of 10 nmol, average mol wt 39,211.28 Da by calculation
A8846 ProteoMass Angiotensin II MALDI-MS Standard, vial of 10 nmol
A8971 ProteoMass Apomyoglobin MALDI-MS Standard, vial of 10 nmol, (M+H+) 16,952.27 Da by calculation
B4181 ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 756.3997 Da
MSCAL1 ProteoMass Peptide and Protein MALDI-MS Calibration Kit, Peptide and Protein MALDI MS Calibration Standard
MSCAL2 ProteoMass Peptide MALDI-MS Calibration Kit, Peptide MALDI MS Calibration Standard
MSCAL3 ProteoMass Protein MALDI-MS Calibration Kit, Protein MALDI MS Calibration Standard
56197 3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
54793 2-(4-Hydroxyphenylazo)benzoic acid, matrix substance for MALDI-MS, ≥99.5%
85429 Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
03565 MALDI validation set polystyrene Mp ~700 - 70 000, certified reference material
37468 2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%
91928 2′,4′,6′-Trihydroxyacetophenone monohydrate, matrix substance for MALDI-MS, ≥99.5%
03597 MALDI validation set PS, PMMA, PDMS, PEG and PSS Mp ~4′300 - 6′500
87884 trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, matrix substance for MALDI-MS, ≥99.0% (HPLC)
60018 Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
29208 (N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide, for protein sequence analysis (by MALDI-MS), ≥98.5%
46278 trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
85707 2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
70990 α-Cyano-4-hydroxycinnamic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
N3786 α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
39468 α-Cyano-4-hydroxycinnamic acid, matrix substance for MALDI-MS, Ultra pure
39319 2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
49508 Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
50862 Super-DHB, suitable for matrix substance for MALDI-MS, ≥99.0%
02241 2′,3′-Dideoxycytidine 5′-triphosphate sodium salt solution, for MALDI MS, ≥98.0% (HPLC)
MSCAL4 ProteoMass MALDI Calibration Kit for Exactive Series MS, LTQ XL and LTQ Hybrids, MALDI MS Calibration Standard
92817 9-Aminoacridine, matrix substance for MALDI-MS, ≥99.5% (HPLC)
56451 1,5-Diaminonaphthalene, matrix substance for MALDI-MS, ≥99.0% (HPLC)
50757 Micro-Array for Mass Spectrometry (MAMS), for single cell analysis
94141 4-Chloro-α-cyanocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
77646 α-Cyano-2,4-difluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
89063 4-Bromo-α-cyanocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
77081 α-Cyano-4-fluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
38419 α-Cyano-2,3,4,5,6-pentafluorocinnamic acid, matrix substance for MALDI-MS, ≥95.0% (HPLC)
39379 4-Chloro-α-cyanocinnamic acid - α-Cyano-2,4-difluorocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of both components, HPLC)
68914 4-Bromo-α-cyanocinnamic acid - 4-Chloro-α-cyanocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of both components, HPLC)
03841 α-Cyano-4-hydroxycinnamic acid - α-Cyano-2,4-difluorocinnamic acid - α-Cyano-2,3,4,5,6-pentafluorocinnamic acid mixture, matrix substance for MALDI-MS, ≥95.0% (sum of three components, HPLC)
05851 4-Aminoquinaldine, matrix substance for MALDI-MS, ≥99.0% (HPLC)
78246 Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
04973 Micro-Array for Mass Spectrometry (MAMS), for quantitative MALDI
94477 (E)-2-Cyano-3-(2-naphthyl)acrylic acid, matrix substance for MALDI-MS, ≥98.0% (HPLC)
83788 (2E)-3-(9-Anthryl)-2-cyanoacrylic acid, matrix substance for MALDI-MS, ≥97.0% (HPLC)
76884 Anthranilamide, matrix substance for MALDI-MS, ≥99.0% (HPLC)
56229 9-Nitroanthracene, matrix substance for MALDI-MS, ≥98.5% (HPLC)
PFS11 SpheriCal Mix, Peptide Low, for MALDI MS
PFS12 SpheriCal Mix, Peptide Medium, for MALDI MS
PFS13 SpheriCal Mix, Peptide High, for MALDI MS
PFS14 SpheriCal Mix, Protein Low, for MALDI MS
PFS15 SpheriCal Mix, Protein Medium, for MALDI MS
PFS31 SpheriCal Aqua, Peptide Medium, for MALDI MS
PFS32 SpheriCal Aqua, Peptide High, for MALDI MS
PFS20 SpheriCal Neat, Peptide Low, for MALDI MS
PFS21 SpheriCal Neat, Peptide Medium, for MALDI MS
PFS22 SpheriCal Neat, Peptide High, for MALDI MS
PFS23 SpheriCal Neat, Protein Low, for MALDI MS
PFS24 SpheriCal Neat, Protein Medium, for MALDI MS
PFS30 SpheriCal Aqua, Peptide Low, for MALDI MS
69028 4-Phenyl-α-cyanocinnamamide, matrix substance for MALDI-MS, ≥98.5% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?