• USA Home
  • PFS64 - SpheriCal Mix CHCA

EMAIL THIS PAGE TO A FRIEND
PFS64 Supelco

SpheriCal Mix CHCA

Protein Low

Purchase

Properties

shelf life   expiry date on the label
composition   α-cyano-4-hydroxycinnamic acid, >80%
  sodium trifluoroacetate, <20%
storage temp.   −20°C

Description

General description

SpheriCal is a dendrimer-based MALDI calibration kit providing 4 monodisperse mass signals per kit for efficient, highly accurate calibration. By combining excellent long-term stability with evenly-spaced calibration points, SpheriCal helps users increase the frequency and quality of their MALDI-MS calibrations. With a shelf life of years and each kit offering at least 100 calibration spots, SpheriCal offers a reliable and accurate calibration across a large mass range, without the shelf-life and storage issues associated with protein or peptide-based systems.

Mass ranges available:

i. SpheriCal Mix DHB Peptide Low (m/z 500 - 1,600)
ii. SpheriCal Mix DHB Peptide Medium (m/z 1,600 - 3,500)
iii. SpheriCal Mix DHB Peptide High (m/z 3,500 - 7,500)
iv. SpheriCal Mix DHB Protein Low (m/z 7,500 - 15,000)
v. SpheriCal DHB Protein Medium (m/z 15,000 - 30,000)

SpheriCal Mix: quick and easy to use - just add solvent

As calibration standard, matrix and counter ion are pre-mixed in optimized ratios, SpheriCal Mix dramatically expedites sample preparation for calibration in minutes. SpheriCal Mix CHCA is pre-formulated with α-cyano-4-hydroxycinnamic acid, a commonly used matrix in the analysis of peptides, proteins, phospholipids, nucleotides and polar synthetic polymers.

Find more information here SpheriCal

dendritic calibrants PFS4A : PFS4B : PFS4C : PFS4D = 1:1:1:1

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Grayson Group - Professor Product Portal

From our library of Articles, Sigma-Aldrich presents Grayson Group - Professor Product Portal
Keywords: Coupling reactions, Mass spectrometry, Nuclear magnetic resonance spectroscopy, Phase transitions, Polymerization reactions, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?