• USA Home
  • PHEN100 - Phenols Total (1000 ug/mL)

EMAIL THIS PAGE TO A FRIEND
PHEN100 Supelco

Phenols Total (1000 ug/mL)

certified reference material

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, M-P, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   certified reference material
mfr. no.   RTC
concentration   in H2O
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   multi-component solution

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Specifications

View example COA for PHEN100

Note - The link provided is an example certificate of analysis (COA) only and may not be the lot that you receive. All analytes may not be present or certified for each lot, and analyte concentrations will vary by lot.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?