• USA Home
  • S4006 - Secobarbital solution

EMAIL THIS PAGE TO A FRIEND
S4006 Supelco

Secobarbital solution

1.0 mg/mL in methanol, analytical standard, for drug analysis

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Barbiturates, Chemical Structure,
grade   analytical standard, for drug analysis
drug control   Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   2-8°C
SMILES string   CCCC(C)C1(CC=C)C(=O)NC(=O)NC1=O
InChI   1S/C12H18N2O3/c1-4-6-8(3)12(7-5-2)9(15)13-11(17)14-10(12)16/h5,8H,2,4,6-7H2,1,3H3,(H2,13,14,15,16,17)
InChI key   KQPKPCNLIDLUMF-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

1 mL in ampule

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Articles

HPLC Analysis of Barbiturates in Serum on Discovery® C18 after SPE using Discovery® DSC-18Lt

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates in Serum on Discovery® C18 after SPE using Discovery® DSC-18Lt
Keywords: Calorimetry, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical, Sample preparations, Separation, Solid phase extractions, Solvents

HPLC Analysis of Barbiturates on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Ascentis® C18
Keywords: Anticonvulsants, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical, Separation, Solvents

HPLC Analysis of Barbiturates on Ascentis® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Ascentis® C8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® C8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® RP-AmideC16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® RP-AmideC16
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?