• USA Home
  • SPE007MA - Diesel in Soil by MA Methods - PT

EMAIL THIS PAGE TO A FRIEND
SPE007MA Supelco

Diesel in Soil by MA Methods - PT

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Solids/Soils Hazardous Waste Organic PTs,
packaging   ampule of 40 g
mfr. no.   RTC, SPE-007MA
matrix   soil
Featured Industry   Environmental
storage temp.   2-8°C

Description

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2018-2019)

Soils
LPTP18-S1 January 31 - March 16, 2018
LPTP18-S2 April 11 - May 25, 2018
LPTP18-S3 July 25 - September 7, 2018
LPTP18-S4 October 10 - November 23, 2018

LPTP19-S1 January 30 - March 15, 2019
LPTP19-S2 April 10 - May 24, 2019
LPTP19-S3 July 24 - September 6, 2019
LPTP19-S4 October 9 - November 22, 2019

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Preparation Note

This sample consists of 10g of soil containing EPH/PAHs in soil. Four samples have been provided for your convenience. The standard may not contain all of the compounds listed on the Report Form. For analytes not detected, report a “<” and your detection limit.

Sample Preparation
Extract the complete contents of a single vial. Transfer entire contents of one vial to extraction vessel.  Rinse vial and cap with extraction solvent.
Note: Sample extracts and calibration solutions should be in the same solvent.
Analyze the extract using the appropriate MA EPH/PAH method.
Determine the concentration of the analytes listed, and report your results on the organic analysis data sheet provided. Report all results on a wet weight basis, do not correct for moisture. Assume a 10g sample size for all calculations.

Analyte

Description

Product #

Add to Cart

Acenaphthene .1-10 mg/kg    
Acenaphthylene .1-10 mg/kg    
Anthracene .1-10 mg/kg    
Benzo(a)anthracene .1-10 mg/kg    
Benzo[a]pyrene .1-10 mg/kg    
Benzo[b]fluoranthene .1-10 mg/kg    
Benzo[ghi]perylene .1-10 mg/kg    
Benzo[k]fluoranthene .1-10 mg/kg    
Chrysene .1-10 mg/kg    
C9-C18 Aliphatic Hydrocarbons .001-4000 mg/kg    
C11-C22 Aromatic Hydrocarbons .001-4000 mg/kg    
C19-C36 Aliphatic Hydrocarbons .001-4000 mg/kg    
Dibenzo(a,h)anthracene .1-10 mg/kg    
Diesel Range Organics (DRO) 300-3000 mg/kg    
Fluoranthene .1-10 mg/kg    
Fluorene .1-10 mg/kg    
Indeno(1,2,3-cd) pyrene .1-10 mg/kg    
2-Methylnaphthalene (β) .1-10 mg/kg    
Naphthalene .1-10 mg/kg    
Phenanthrene .1-10 mg/kg    
Pyrene .1-10 mg/kg    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products.Once you log onto our website, you will find the price and availability displayed on the product detail page.You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 orview local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Are Certificates of Analysis created forRTC proficiency testing products?
Certificates of Analysis are not available for RTC proficiency testing products. Those are the samples with the SPE or PE prefix in the part number.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SQC007MA Diesel in Soil by MA Methods, certified reference material
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?