• USA Home
  • SPE026TXL - TPH in Soil - Low - PT

EMAIL THIS PAGE TO A FRIEND
SPE026TXL Supelco

TPH in Soil - Low - PT

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Solids/Soils Hazardous Waste Organic PTs,
packaging   ampule of 1 × 30 g
mfr. no.   RTC, SPE-026TX-L
concentration   in pentane
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

The prepared PT sample is a soil containing petroleum hydrocarbons (TPH). The sample size provided is 60g of soil. The soil has been sterilized to minimize degradation of the sample.

Sample Preparation
This is a two part sample including a fortification ampule and a blank soil.
To prepare, weight 10.0 grams of blank soil and transfer to your extraction vessel.
Open ampule SPE-026TX-L and transfer 1.0mL of concentration using a Class A pipet to your extraction vessel. The liquid should be applied directly to the soil this will minimize evaporation losses.
Analyze using Method TX 1005 or TX 1006.
A modified report form can be obtained for method TX 1006 by contacting RTC.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2019-2020)

Soils
LPTP19-S1 January 30 - March 15, 2019
LPTP19-S2 April 10 - May 24, 2019
LPTP19-S3 July 24 - September 6, 2019
LPTP19-S4 October 9 - November 22, 2019

LPTP20-S1 January 29 - March 13, 2020
LPTP20-S2 April 8 - May 22, 2020
LPTP20-S3 July 22 - September 4, 2020
LPTP20-S4 October 7 - November 20, 2020

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Analyte

Description

Product #

Add to Cart

C6 Aliphatics .001-100 %    
>C6 to C8 Aliphatics .001-100 mg/kg    
>C8 to C10 Aliphatics .001-100 mg/kg    
>C10 to C12 Aliphatics .001-100 mg/kg    
>C12 to C16 Aliphatics .001-100 mg/kg    
>C16 to C21 Aliphatics .001-100 mg/kg    
>C21 to C35 Aliphatics .001-100 mg/kg    
>C7-C8 Aromatics .001-100 %    
>C8 to C10 Aromatics .001-100 mg/kg    
>C10 to C12 Aromatics .001-100 mg/kg    
>C12 to C16 Aromatics .001-100 mg/kg    
>C16 to C21 Aromatics .001-100 mg/kg    
>C21 to C35 Aromatics .001-100 mg/kg    
Diesel-range total petroleum hydrocarbons, >C12-C28 25-100 mg/kg    
Gasoline Range Organics, C6-C12 25-100 mg/kg    
Residual Range Organics (RRO),>C28-C35 25-100 mg/kg    
Total Petroleum Hydrocarbons (TPH), (C6-C35) 50-300 mg/kg    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 2 - Pentanes solution
WGK Germany 
3
Flash Point(F) 
-56.2 °F
Flash Point(C) 
-49 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products.Once you log onto our website, you will find the price and availability displayed on the product detail page.You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 orview local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Are Certificates of Analysis created forRTC proficiency testing products?
Certificates of Analysis are not available for RTC proficiency testing products. Those are the samples with the SPE or PE prefix in the part number.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?