• USA Home
  • SPE028 - Toxaphene in Soil - PT

EMAIL THIS PAGE TO A FRIEND
SPE028 Supelco

Toxaphene in Soil - PT

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Solids/Soils Hazardous Waste Organic PTs,
packaging   ampule of 1 × 50 g
mfr. no.   RTC, SPE-028
matrix   soil
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2018-2019)

Soils
LPTP18-S1 January 31 - March 16, 2018
LPTP18-S2 April 11 - May 25, 2018
LPTP18-S3 July 25 - September 7, 2018
LPTP18-S4 October 10 - November 23, 2018

LPTP19-S1 January 30 - March 15, 2019
LPTP19-S2 April 10 - May 24, 2019
LPTP19-S3 July 24 - September 6, 2019
LPTP19-S4 October 9 - November 22, 2019

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Preparation Note

The PT sample size provided is 50 g of soil. The soil has been sterilized to minimize degradation of the sample. The PT sample has been sized to 100 mesh. Required storage condition is 4ºC. The evaluation of results will be based on study statistics as required by NELAC. The sample has been intentionally prepared with an apparent headspace.

Sample Preparation
Mix well prior to subsampling.
Analyze the sample using the preparatory and determinative method(s) for which you are seeking accreditation.
If you are accrediting for more than one method make additional copies of the report form and clearly indicate the method used for the data being reported.
Determine the concentration and report your results on the analysis data sheet provided.
Determine moisture content of the soil and report on a dry weight basis.

Analyte

Description

Product #

Add to Cart

Toxaphene (Chlorinated camphene) 200-2000 μg/kg    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SQC028 Toxaphene in Soil, certified reference material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?