• USA Home
  • SPE068 - PCB Congeners in Soil - PT

EMAIL THIS PAGE TO A FRIEND
SPE068 Supelco

PCB Congeners in Soil - PT

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Organic Pollutant Standards, PCBs & Dioxins Standards,
packaging   ampule of 1 × 50 g
mfr. no.   RTC, SPE-068
matrix   soil
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2018-2019)

Soils
LPTP18-S1 January 31 - March 16, 2018
LPTP18-S2 April 11 - May 25, 2018
LPTP18-S3 July 25 - September 7, 2018
LPTP18-S4 October 10 - November 23, 2018

LPTP19-S1 January 30 - March 15, 2019
LPTP19-S2 April 10 - May 24, 2019
LPTP19-S3 July 24 - September 6, 2019
LPTP19-S4 October 9 - November 22, 2019

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Preparation Note

The PT sample size provided is 50 g of soil. The soil has been sterilized to minimize degradation of the sample. The PT sample has been sized to 100 mesh.
The sample has been intentionally prepared with an apparent headspace.

Sample Preparation
Mix Prior to use.
Recommended minimum sampling size is 1 gram.
Analyze The sample using the preparatory and determinative method(s) for which you are seeking accreditation.
Note: Sample extracts and calibration solutions should be in the same solvent. Report on a dry weight basis.

Analyte

Description

Product #

Add to Cart

2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB 180) 5-500 μg/kg    
2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB 189) 10-500 μg/kg    
2,2',3,4,4',5'-Hexachlorobiphenyl (PCB 138) 5-500 μg/kg    
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB 153) 5-500 μg/kg    
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB 157) 10-500 μg/kg    
2,3,3',4,4',5-Hexachlorobiphenyl (PCB 156) 10-500 μg/kg    
2,3',4,4',5,5'-Hexachlorobiphenyl (PCB 167) 10-500 μg/kg    
3,3',4,4',5,5'-Hexachlorobiphenyl (PCB 169) 10-500 μg/kg    
2,2',4,5,5'-Pentachlorobiphenyl (PCB 101) 5-500 μg/kg    
2,3,3',4,4'-Pentachlorobiphenyl (PCB 105) 10-500 μg/kg    
2,3',4,4',5-Pentachlorobiphenyl (PCB 118) 5-500 μg/kg    
2,3',4,4',5'-Pentachlorobiphenyl (PCB 123) 10-500 μg/kg    
2,3,4,4',5-Pentachlorobiphenyl (PCB 114) 10-500 μg/kg    
3,3',4,4',5-Pentachlorobiphenyl (PCB 126) 10-500 μg/kg    
2,2',5,5'-Tetrachlorobiphenyl (PCB 52) 5-500 μg/kg    
3,3',4,4'-Tetrachlorobiphenyl (PCB 77) 10-500 μg/kg    
3,4,4',5-Tetrachlorobiphenyl (PCB 81) 10-500 μg/kg    
2,4,4'-Trichlorobiphenyl (PCB 28) 5-500 μg/kg    
PCB (20)+(28) .001-1000 μg/kg    
PCB (90)+(101)+(113) .001-1000 μg/kg    
PCB (129)+(138)+(163) .001-1000 μg/kg    
PCB (153)+(168) .001-1000 μg/kg    
PCB (156)+(157) 10-1000 μg/kg    
PCB (180)+(193) .001-1000 μg/kg    
PCBs, total 5-5000 μg/kg    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SPE068 PCB Congeners in Soil - PT, Proficiency Testing Material
SQC068 PCB Congeners in Soil, certified reference material

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?