EMAIL THIS PAGE TO A FRIEND
SPE070 Supelco

Solvent Screen - PT

Proficiency Testing Material

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Proficiency Testing, Solids/Soils Hazardous Waste Organic PTs,
packaging   ampule of 1 × 20 mL
mfr. no.   RTC, SPE-070
concentration   in methanol
Featured Industry   Environmental

Description

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2018-2019)

Soils
LPTP18-S1 January 31 - March 16, 2018
LPTP18-S2 April 11 - May 25, 2018
LPTP18-S3 July 25 - September 7, 2018
LPTP18-S4 October 10 - November 23, 2018

LPTP19-S1 January 30 - March 15, 2019
LPTP19-S2 April 10 - May 24, 2019
LPTP19-S3 July 24 - September 6, 2019
LPTP19-S4 October 9 - November 22, 2019

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)

Preparation Note

This PT sample is packaged as a 20.0 mL whole-volume ampule.
The solvent for this sample is methanol.
The PT sample is not preserved.
This sample may or may not contain all analytes listed on the report form. You are not required to report all analytes only analytes that you wish to be evaluated.

Sample Preparation
The sample in the ampule is ready for analysis.
Dilute as required by your method, but report as received.

Analyte

Description

Product #

Add to Cart

Acetone .1-20 wt. %    
2-Butanone (Methyl ethyl ketone, MEK) .05-5 wt. %    
tert-Butanol solution .025-20 wt. %    
n-Butyl alcohol .025-20 wt. %    
Chloroform .025-1 wt. %    
Dichloromethane (DCM, Methylene chloride) .025-1 wt. %    
Ethanol denatured .025-20 wt. %    
Isopropyl alcohol (2-Propanol) .025-20 wt. %    
Methanol .5-100 wt. %    
4-Methyl-2-pentanone (MIBK) .05-20 wt. %    
Tetrahydrofuran (THF) .05-20 wt. %    
Toluene .025-5 wt. %    
m+p-Xylene .05-20 wt. %    
m-Xylene .5-20 wt. %    
o-Xylene .05-20 wt. %    
p-Xylene .5-20 wt. %    
Xylene, total .05-20 wt. %    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1992 6.1(3) / PGII
WGK Germany 
2
Flash Point(F) 
68 °F
Flash Point(C) 
20 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SQC070 Solvent Screen, certified reference material
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?