EMAIL THIS PAGE TO A FRIEND
UST1081 Supelco

EPH by GC/FID

certified reference material, 50000 μg/mL in methylene chloride

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Environmental Matrix CRMs (RTC),
grade   certified reference material
packaging   pkg of 1.5 mL
mfr. no.   RTC, UST-108-1
concentration   50000 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
volume size   1.5 mL
Featured Industry   Environmental
Petroleum
format   matrix material

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

Product #

Add to Cart

30W Motor Oil 25 wt. %    
Diesel Fuel #2 25 wt. %    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?