• USA Home
  • UST116 - EPH Aromatic Hydrocarbon Standard

EMAIL THIS PAGE TO A FRIEND
UST116 Supelco

EPH Aromatic Hydrocarbon Standard

certified reference material, 1000 μg/mL in methylene chloride

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Environmental Matrix CRMs (RTC),
grade   certified reference material
packaging   pkg of 1.5 mL
mfr. no.   RTC, UST-116
concentration   1000 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
Petroleum
format   matrix material

Description

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

Product #

Add to Cart

Decane    
Dodecane    
Heneicosane    
Hexadecane    
Octane    
Tetratriacontane    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?