• USA Home
  • UST133 - TNRCC Method 1005 Retention Time Marker

EMAIL THIS PAGE TO A FRIEND
UST133 Supelco

TNRCC Method 1005 Retention Time Marker

certified reference material, 500 μg/mL in pentane

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs), Environmental Matrix CRMs (RTC),
grade   certified reference material
packaging   pkg of 1.5 mL
mfr. no.   RTC, UST-133
concentration   500 μg/mL in pentane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
Petroleum
format   matrix material

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

Product #

Add to Cart

Decane    
Dodecane    
Heneicosane    
Heptane    
Hexadecane    
Hexane    
Octacosane    
Octadecane    
Octane    
Pentatriacontane    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 2 - Pentanes solution
WGK Germany 
2
Flash Point(F) 
-56.2 °F
Flash Point(C) 
-49 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?