• USA Home
  • VT001077 - Legionella pneumophila WDCM 00107 Vitroids

EMAIL THIS PAGE TO A FRIEND
VT001077 Supelco

Legionella pneumophila WDCM 00107 Vitroids

≥ 100′000CFU mean value range, certified reference material, for microbiology

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Bacteria Standards, Microbiological Standards, Microbiology,
Quality Level   300
grade   certified reference material
  for microbiology
Featured Industry   Environmental
Food and Beverages
storage temp.   −20°C
suitability   Legionella spp.

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Other Notes

ATCC® 33152, NCTC 11192, NCPF -, DSM 7513, LMG -, IHEM -, CIP 103854T

Origin Strain No. CECT® 7109

Legal Information

Derived from CECT® strains

ATCC is a registered trademark of American Type Culture Collection

CECT is a registered trademark of Universitat de Valencia

Vitroids is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
UN3373 - class 6.2 Biological substance, Category B
WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

USDA Interstate Transport Permit Process for Vitroids™ and Lenticule® Discs

Customers in the United States, who are located outside of Missouri (where the products are distributed), should apply for and obtain a USDA Transport Permit to receive Vitroids™ and Lenticule® Disc ...
Keywords: Cell culture, Veterinary

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?