• USA Home
  • W559001 - 1-(2-Thienyl)-1-propanone

EMAIL THIS PAGE TO A FRIEND
W559001 Sigma-Aldrich

1-(2-Thienyl)-1-propanone

≥97%, FG

Synonym: 2-Propionylthiophene

Purchase

Properties

Related Categories Alphabetical Listings, Certified Food Grade Products, Flavors and Fragrances, Halal Certified Products, Kosher Certified Products,
biological source   synthetic
grade   FG
  Halal
  Kosher
reg. compliance   EU Regulation 1334/2008 & 178/2002
InChI Key   MFPZQZZWAMAHOY-UHFFFAOYSA-N
assay   ≥97%
refractive index   n20/D 1.553(lit.)
bp   107 °C/11 mmHg(lit.)
density   1.126 g/mL at 25 °C(lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   caramel; creamy
food allergen   no known allergens

Description

Packaging

25, 100 g in poly bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?