• USA Home
  • Y0000623 - Terazosin impurity L

EMAIL THIS PAGE TO A FRIEND
Y0000623 Sigma-Aldrich

Terazosin impurity L

European Pharmacopoeia (EP) Reference Standard

Synonym: 1-(2-Furoyl)piperazine, 2-Furoic piperazide

  • CAS Number 40172-95-0

  • Empirical Formula (Hill Notation) C9H12N2O2

  • Molecular Weight 180.20

  •  Beilstein/REAXYS Number 880196

  •  MDL number MFCD00038831

  •  PubChem Substance ID 329831058

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, EP Standards, EP Standards S - T, Pharmacopeia & Metrological Institutes Standards More...
grade   pharmaceutical primary standard
InChI Key   SADPINFEWFPMEA-UHFFFAOYSA-N
form   neat
mp   67-70 °C (lit.)
Featured Industry   Pharmaceutical (small molecule)
format   neat

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference subtance catalogue.

Caution

Please find SDS provided by EDQM here..

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?