• USA Home
  • Y0000625 - Vecuronium for peak identification

EMAIL THIS PAGE TO A FRIEND
Y0000625

Vecuronium for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym: 1-[(2β,3α,5α,16β,17β)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methyl-piperidinium bromide, Vecuronium bromide

  • CAS Number 50700-72-6

  • Empirical Formula (Hill Notation) C34H57BrN2O4

  • Molecular Weight 637.73

  •  Beilstein/REAXYS Number 4837308

  •  MDL number MFCD00867762

  •  PubChem Substance ID 329831060

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, EP Standards, EP Standards U - Z, Pharmacopeia & Metrological Institutes Standards More...
form   neat
Featured Industry   Pharmaceutical (small molecule)
format   neat
SMILES string   [Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(C)=O)[N+]5(C)CCCCC5)[C@@]2(C)C[C@@H]1N6CCCCC6
InChI   1S/C34H57N2O4.BrH/c1-23(37)39-31-20-25-12-13-26-27(34(25,4)22-29(31)35-16-8-6-9-17-35)14-15-33(3)28(26)21-30(32(33)40-24(2)38)36(5)18-10-7-11-19-36;/h25-32H,6-22H2,1-5H3;1H/q+1;/p-1/t25-,26+,27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1
InChI key   VEPSYABRBFXYIB-PWXDFCLTSA-M
Gene Information   human ... CHRNA1(1134), CHRNB1(1140), CHRND(1144), CHRNE(1145), CHRNG(1146)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference subtance catalogue.

Caution

Please find SDS provided by EDQM here..

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?