• USA Home
  • Y0000629 - Enalaprilat for system suitability

EMAIL THIS PAGE TO A FRIEND
Y0000629

Enalaprilat for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym: (2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenyl-propyl]amino]propanoyl]pyrrolidine-2-carboxylic acid, Enalaprilat dihydrate, Vasotec

  • CAS Number 84680-54-6

  • Empirical Formula (Hill Notation) C18H24N2O5 ·2H2O

  • Molecular Weight 384.42

  •  MDL number MFCD00941393

  •  PubChem Substance ID 329831062

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, EP Standards, EP Standards E - F, Pharmacopeia & Metrological Institutes Standards More...
form   neat
Featured Industry   Pharmaceutical (small molecule)
format   neat
storage temp.   2-8°C
SMILES string   O.O.C[C@H](N[C@@H](CCc1ccccc1)C(O)=O)C(=O)N2CCC[C@H]2C(O)=O
InChI   1S/C18H24N2O5.2H2O/c1-12(16(21)20-11-5-8-15(20)18(24)25)19-14(17(22)23)10-9-13-6-3-2-4-7-13;;/h2-4,6-7,12,14-15,19H,5,8-11H2,1H3,(H,22,23)(H,24,25);2*1H2/t12-,14-,15-;;/m0../s1
InChI key   MZYVOFLIPYDBGD-MLZQUWKJSA-N
Gene Information   human ... ACE(1636)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Enalaprilat is an inhibitor of angiotensin converting enzyme (ACE), antihypertensive, and a Bradykinin B1 receptor activator. Enalaprilat has nM potency versus ACE and also activates B1 receptors to release NO.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Caution

Please find SDS provided by EDQM here..

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?