• USA Home
  • Y0001478 - Thiopental for system suitability

EMAIL THIS PAGE TO A FRIEND
Y0001478

Thiopental for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym: (±)-Thiopental, 5-Ethyl-5-(1-methylbutyl)-2-thiobarbituric acid, Pentothal

  • CAS Number 76-75-5

  • Empirical Formula (Hill Notation) C11H18N2O2S

  • Molecular Weight 242.34

  •  MDL number MFCD00057564

  •  PubChem Substance ID 329831446

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, EP Standards, EP Standards S - T, Pharmacopeia & Metrological Institutes Standards More...
grade   pharmaceutical primary standard
form   neat
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
Featured Industry   Pharmaceutical (small molecule)
format   neat
SMILES string   CCCC(C)C1(CC)C(=O)NC(=S)NC1=O
InChI   1S/C11H18N2O2S/c1-4-6-7(3)11(5-2)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)
InChI key   IUJDSEJGGMCXSG-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Caution

Please find SDS provided by EDQM here..

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?