• USA Home
  • Y0001602 - Methylprednisolone for system suitability

EMAIL THIS PAGE TO A FRIEND
Y0001602

Methylprednisolone for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym: 11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 6α-Methylprednisolone, Medrol, Medrone

  • CAS Number 83-43-2

  • Empirical Formula (Hill Notation) C22H30O5

  • Molecular Weight 374.47

  •  Beilstein/REAXYS Number 2340300

  •  MDL number MFCD00010591

  •  PubChem Substance ID 329831507

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, EP Standards, EP Standards L - M, Pharmacopeia & Metrological Institutes Standards More...
grade   pharmaceutical primary standard
form   neat
mfr. no.   EDQM
Featured Industry   Pharmaceutical (small molecule)
format   neat
SMILES string   [H][C@@]12C[C@H](C)C3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO
InChI   1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1
InChI key   VHRSUDSXCMQTMA-PJHHCJLFSA-N
Gene Information   human ... NR3C1(2908)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Caution

Please find SDS provided by EDQM here..

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?