• USA Home
  • Z801917 - Hellma® liquid calibration standards

EMAIL THIS PAGE TO A FRIEND
Z801917

Hellma® liquid calibration standards

UV100, for testing the photometric accuracy

  •  NACRES SB.32

Purchase

Description

General description

Certified liquid Hellma calibration standard to enable the checking of spectrophotometers with respect to photometric accuracy.

Consisting of :
100 mg Potassium dichromate in perchloric acid (667-UV100, wavelength 235; 257; 313; 350 nm)

Other Notes

The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.

Legal Information

Hellma is a registered trademark of Hellma GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?