• USA Home
  • Z802506 - Hellma® absorption cuvettes, Flow-through cells
EMAIL THIS PAGE TO A FRIEND
Z802506
Hellma® absorption cuvettes, Flow-through cells
optical glass, spectral range 320-2500 nm, pathlength 10 mm, chamber volume 750 μL
  •  NACRES SB.32
Purchase
Properties
material   optical glass
mfr. no.   Hellma 175.000-OS
parameter   320-2500 nm spectral range
application(s)   UV/Vis spectroscopy: suitable
H × W × D   38.5 mm × 12.5 mm × 7.5 mm
aperture center H   8.5 mm
chamber volume   750 μL
pathlength   10 mm
Description
General description
2 polished windows; Macro, with in/outlet tubes.
2 windows polished aperture.
Legal Information
Hellma is a registered trademark of Hellma GmbH
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?