• USA Home
  • Z802948 - Hellma® absorption cuvettes, Flow-through cells
EMAIL THIS PAGE TO A FRIEND
Z802948
Hellma® absorption cuvettes, Flow-through cells
Suprasil® quartz, spectral range 200-2500 nm, pathlength 10 mm, chamber volume 450 μL
  •  NACRES SB.32
Purchase
Properties
material   Suprasil® quartz
mfr. no.   Hellma 176.000-QS
parameter   200-2500 nm spectral range
application(s)   UV/Vis spectroscopy: suitable
H × W × D   45 mm × 12.5 mm × 12.5 mm
aperture center H   15 mm
chamber volume   450 μL
pathlength   10 mm
Description
General description
2 polished windows; Semi-micro, with in/outlet tubes
2 windows polished aperture.
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?