• USA Home
  • Z803014 - Hellma® TrayCell cap

EMAIL THIS PAGE TO A FRIEND
Z803014

Hellma® TrayCell cap

pathlength 2 (factor 5) mm ± 0.02 mm, chamber volume 6-10 μL

  •  NACRES SB.32

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Cuvettes, Cuvettes for Special Application, Hellma,
mfr. no.   Hellma (665.705)
application(s)   UV/Vis spectroscopy: suitable
  photometry: suitable
chamber volume   6-10 μL
pathlength   2 (factor 5) mm ± 0.02 mm

Description

General description

Cap made of stainless steel with a mirror made of Quartz Suprasil® with an aluminum mirror layer.
factor = dilution factor compared to a standard cell with a path length of 10 mm

Legal Information

Hellma is a registered trademark of Hellma GmbH

Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.

TrayCell is a trademark of Hellma GmbH

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

Z802573 Hellma® TrayCell, Suprasil® quartz, spectral range 200-2500 nm, pathlength 0.2 (factor 50) mm ± 0.02 mm × 1.0 (factor 10) mm ± 0.02 mm, chamber volume 0.7-4 μL
Z802697 Hellma® TrayCell, Suprasil® quartz, spectral range 200-2500 nm, pathlength 0.2 (factor 50) mm ± 0.02 mm × 1.0 (factor 10) mm ± 0.02 mm, chamber volume 0.7-4 μL
Z802905 Hellma® TrayCell cap, pathlength 0.2 (factor 50) mm ± 0.02 mm, chamber volume 0.7-4 μL
Z802808 Hellma® TrayCell cap, pathlength 1 (factor 10) mm ± 0.02 mm, chamber volume 3-5 μL
Z803138 Hellma® TrayCell cap, pathlength 0.1 (factor 100) mm ± 0.02 mm, chamber volume 0.7-3 μL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?